Anti-UPK3A Rabbit Polyclonal Antibody (CF405S) [clone: Rabbit PAb]

Supplier: Biotium

BNC040409-100 BNC040409-500
75980-564EA 325.12 CAD
75980-564 75980-942
Anti-UPK3A Rabbit Polyclonal Antibody (CF405S) [clone: Rabbit PAb]
Uroplakin-3A (UPK3A) is a component of the asymmetric unit membrane (AUM). It is a highly specialized bio-membrane made by terminally differentiated urothelial cells. The protein may play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. UPK3A also contributes to the formation of urothelial glycocalyx, which may play an important role in preventing bacterial adherence through FimH bacterial protein binding leading to bladder infection. UPK3A has been shown as a helpful marker for the detection of bladder cancer.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: UPK3A
Clonality: Polyclonal
Clone: Clone Rabbit PAb
Conjugation: CF405S
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR