Anti-TIMP3 Rabbit Polyclonal Antibody (CF594) [clone: Rabbit PAb]

Supplier: Biotium

BNC940408-100 BNC940408-500
75980-890EA 325.12 CAD
75980-890 75980-892
Anti-TIMP3 Rabbit Polyclonal Antibody (CF594) [clone: Rabbit PAb]
Antibodies
TIMP3 (tissue inhibitor of metalloproteinases 3), along with family members TIMP1, TIMP2, and TIMP4, are inhibitors of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix (ECM). An imbalance between MMPs and the associated TIMPs may play a significant role in the invasive phenotype of malignant tumors. TIMP s inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells. TIMP-3 may be involved in regulating trophoblastic invasion of the uterus as well as in regulating remodeling of the extracellular matrix during the folding of epithelia, and in the formation, branching and expansion of epithelial tubes.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: TIMP3
Clonality: Polyclonal
Clone: Clone Rabbit PAb
Conjugation: CF594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR