Anti-ESR1 Mouse Monoclonal Antibody (CF405S) [clone: AER314]

Supplier: Biotium

BNC040406-100 BNC040406-500
75924-828EA 325.12 CAD
75924-828 75924-830
Anti-ESR1 Mouse Monoclonal Antibody (CF405S) [clone: AER314]
Antibodies
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa120-170. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: Clone AER314
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow, Human, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR