Anti-Cytokeratins Mouse Monoclonal Antibody (CF594) [clone: C11]

Supplier: Biotium

BNC940393-500 BNC940393-100
75916-950EA 900.17 CAD
75916-950 75918-230
Anti-Cytokeratins Mouse Monoclonal Antibody (CF594) [clone: C11]
Antibodies
This MAb recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. This is a broad-spectrum antibody, which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: Multiple
Clonality: Monoclonal
Clone: Clone C11
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow, Frog, Goat, Guinea Pig, Human, Marmoset, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR