Anti-Cytokeratins Mouse Monoclonal Antibody (CF555) [clone: C11]

Supplier: Biotium

BNC550393-100 BNC550393-500
75916-028EA 325.12 CAD
75916-028 75916-030
Anti-Cytokeratins Mouse Monoclonal Antibody (CF555) [clone: C11]
Antibodies
This MAb recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. This is a broad-spectrum antibody, which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®555 is a bright orange-red fluorescent dye (Ex/Em 555/565 nm) that is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: Multiple
Clonality: Monoclonal
Clone: Clone C11
Conjugation: CF555
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow, Frog, Goat, Guinea Pig, Human, Marmoset, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR