Anti-TNC Mouse Monoclonal Antibody (Biotin) [clone: T2H5]

Supplier: Biotium

BNCB0392-100 BNCB0392-500
75975-976EA 325.12 CAD
75975-976 75975-978
Anti-TNC Mouse Monoclonal Antibody (Biotin) [clone: T2H5]
Antibodies
In Western blotting, this antibody reacts with two bands of ~MW of 210 kDa and 300 kDa, identified as two isoforms of Tenascin C. Specificity of this MAb is validated by sequential immunoprecipitation with a PAb against Tenascin C. Tenascin C is a multifunctional, disulfide-linkedhexameric extracellular matrix glycoprotein expressed in association with mesenchymal epithelial interactions during development and in the neo-vasculature and stroma of undifferentiated tumors. In adults, it is restricted to certain epithelial-stromal interfaces and increases markedly in hyper-proliferative diseases and in stroma of many neoplasms, including gliomas, breast, squamous and lung carcinomas.

Type: Primary
Antigen: TNC
Clonality: Monoclonal
Clone: Clone T2H5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR