Anti-Helicobacter pylori Rabbit Polyclonal Antibody (CF647)

Supplier: Biotium

BNC470374-100 BNC470374-500
75980-642EA 325.12 CAD
75980-642 75980-644
Anti-Helicobacter pylori Rabbit Polyclonal Antibody (CF647)
Antibodies
The spiral shaped bacterium Helicobacter pyloriis strongly associated with inflammation of the stomach and is also implicated in the development of gastric malignancy. H. pylori is known to cause peptic ulcers and chronic gastritis in humans. It is associated with duodenal ulcers and may be involved in development of adenocarcinoma and low-grade lymphoma of mucosa associated lymphoid tissue in the stomach. This antibody stains the individual H. pylori bacterium when it presents on the surface of the epithelium or in the cytoplasm of the epithelial cells in biopsy tissue sections from the antrum and body of the stomach.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: Helicobacter pylori
Clonality: Polyclonal
Clone:
Conjugation: CF647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Helicobacter pylori
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR