Anti-Helicobacter pylori Rabbit Polyclonal Antibody (CF640R)

Supplier: Biotium

BNC400374-100 BNC400374-500
75980-638EA 325.12 CAD
75980-638 75980-640
Anti-Helicobacter pylori Rabbit Polyclonal Antibody (CF640R)
Antibodies
The spiral shaped bacterium Helicobacter pyloriis strongly associated with inflammation of the stomach and is also implicated in the development of gastric malignancy. H. pylori is known to cause peptic ulcers and chronic gastritis in humans. It is associated with duodenal ulcers and may be involved in development of adenocarcinoma and low-grade lymphoma of mucosa associated lymphoid tissue in the stomach. This antibody stains the individual H. pylori bacterium when it presents on the surface of the epithelium or in the cytoplasm of the epithelial cells in biopsy tissue sections from the antrum and body of the stomach.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: Helicobacter pylori
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Helicobacter pylori
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR