Anti-CD6 Mouse Monoclonal Antibody (CF568) [clone: C6/372]

Supplier: Biotium

BNC680372-100 BNC680372-500
75917-166EA 325.12 CAD
75917-166 75917-168
Anti-CD6 Mouse Monoclonal Antibody (CF568) [clone: C6/372]
Antibodies
CD6 is a type I transmembrane glycoprotein that contains a 24-amino acid signal sequence, three extracellularscavenger receptor cysteine-rich(SRCR) domains, a membrane-spanning domain and a 44-amino acid cytoplasmic domain. The CD6 glycoprotein is tyrosine phosphorylated during TCR-mediated T cell activation. CD6 shows significant homology to CD5. CD6 is present on mature thymocytes, peripheral T cells and a subset of B cells. Antibodies to CD6 are used to deplete T cells from bone marrow transplants to prevent graft versus host disease.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: Clone C6/372
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR