Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (Biotin) [clone: AE-1 / AE-3]

Supplier: Biotium

BNCB0371-100 BNCB0371-500
75911-138EA 325.12 CAD
75911-138 75911-140
Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (Biotin) [clone: AE-1 / AE-3]
Antibodies
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, which 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8); 56.5 kDa (CK10); 50 kDa (CK14); 50 kDa (CK15); 48 kDa (CK16); 40 kDa (CK19). Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. AE-1/AE-3 is a broad spectrum anti pan-cytokeratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer. It has been used to characterize the source of various neoplasms and to study the distribution of cytokeratin containing cells in epithelia during normal development and during the development of epithelial neoplasms. This antibody stains cytokeratins present in normal and abnormal human tissues and has shown high sensitivity in the recognition of epithelial cells and carcinomas.

Type: Primary
Antigen: KRT77 & KRT76
Clonality: Monoclonal
Clone: Clone AE-1 / AE-3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG's
Reactivity: Chicken, Cow, Dog, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR