Anti-BSG Mouse Monoclonal Antibody [clone: CB43]

Supplier: Biotium

BNUB0363-100 BNUB0363-500
75887-764EA 380.65 CAD
75887-764 75887-766
Anti-BSG Mouse Monoclonal Antibody [clone: CB43]
Antibodies
CD147 is a transmembrane glycoprotein of the immunoglobulin superfamily. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is involved in the regulation of matrix remodeling at the epidermal-dermal interface. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes, which are typically increased during tissue degradation and wound healing, are important factors in cancer invasion and metastasis.

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: Clone CB43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR