Anti-Histiocytoma Marker Mouse Monoclonal Antibody (Biotin) [clone: D11]

Supplier: Biotium

BNCB0348-100 BNCB0348-500
75952-924EA 325.12 CAD
75952-924 75952-926
Anti-Histiocytoma Marker Mouse Monoclonal Antibody (Biotin) [clone: D11]
Antibodies
In Western blotting, this antibody detects an antigen of 125 kDa in human liver and 135 kDa in tumors of histiocytic origin. Comparative study of this MAb and a standard CD68 MAb showed that their antigens are different. Its antigen in all macrophage types studied is located on the plasma membrane and within cytoplasmic structures including lysosomes. This MAb shows a restricted reactivity to cells of the monocyte/macrophage system. It specifically reacts with blood monocytes and stains resident macrophages in a wide variety of human tissues. This MAb does not stain antigen-presenting cells, e.g., Langerhans cells. Reportedly, its reactivity is restricted to histiocytes and macrophages.

Type: Primary
Antigen: Histiocytoma Marker
Clonality: Monoclonal
Clone: Clone D11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR