Anti-SPN Mouse Monoclonal Antibody (CF488A) [clone: 84-3C1]
Supplier: Biotium
BNC880342-100 BNC880342-500
75895-822EA 325.12 CAD
75895-822 75895-824
Anti-SPN Mouse Monoclonal Antibody (CF488A) [clone: 84-3C1]
Antibodies
This antibody recognizes a cell surface glycoprotein of 95/115/135 kDa (depending upon the extent of glycosylation), identified as CD43 (Workshop III). 70-90% of T-cell lymphomas and from 22-37% of B-cell lymphomas express CD43. No reactivity has been observed with reactive B-cells. So a B-lineage population that co-expresses CD43 is highly likely to be a malignant lymphoma, especially a low-grade lymphoma, rather than a reactive B-cell population. When CD43 antibody is used in combination with anti-CD20, effective immunophenotyping of the lymphomas in formalin-fixed tissues can be obtained.More Product Information