Anti-CD8A Mouse Monoclonal Antibody (CF488A) [clone: RIV11]
Supplier: Biotium
BNC880333-100 BNC880333-500
75907-232EA 325.12 CAD
75907-232 75907-234
Anti-CD8A Mouse Monoclonal Antibody (CF488A) [clone: RIV11]
Antibodies
This antibody recognizes a protein of 32 kDa, identified as CD8a (also known as CD8 chain, T cell co-receptor, Leu2, and T8). CD8 molecule consists of two chains, termed and chain, which are expressed as a disulphide-linked heterodimer or as an homodimer. CD8 is expressed on T cell subset (cytotoxic/suppressor T cells), thymocytes and NK cells. The majority of CD8 T-cells expresses CD8 as heterodimer. Some subpopulation of CD8 T cells as well as NK cells may express homodimer. CD8 functions as a co-receptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelopeMore Product Information