Anti-CD3E Mouse Monoclonal Antibody (CF568) [clone: RIV9]

Supplier: Biotium

BNC680322-100 BNC680322-500
75895-490EA 325.12 CAD
75895-490 75895-492
Anti-CD3E Mouse Monoclonal Antibody (CF568) [clone: RIV9]
Antibodies
This antibody reacts with five invariable CD3 chains designated CD3y, CD3d, CD3e, CD3f, and CD3n with molecular weight ranging from 16-28 kDa. It is found on all mature T-lymphocytes, NK, T-cells, and some thymocytes. CD3, also known as T3, is a member of the immunoglobulin superfamily that plays a role in antigen recognition, signal transduction, and T cell activation. It is expressed, typically at high levels, on peripheral T cells and majority of T cell neoplasms. Thymocytes express CD3 at different level on the cell surface in the course of differentiation and, in cortical thymus, CD3 is predominantly intracytoplasmic. This MAb is particularly useful for induction of T cell activation in vitro and in vivo due to its unusual IgG3 isotype.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: Clone RIV9
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR