Anti-ITGB2 Mouse Monoclonal Antibody (CF405S) [clone: 68-5A5]

Supplier: Biotium

BNC040321-100 BNC040321-500
75888-894EA 325.12 CAD
75888-894 75888-896
Anti-ITGB2 Mouse Monoclonal Antibody (CF405S) [clone: 68-5A5]
Antibodies
This antibody recognizes a transmembrane glycoprotein of 95 kDa, identified as CD18 or integrin -2 (Workshop III). It complexes non-covalently with either L, M, or X integrin (CD11a, b, or c) to form the heterodimers, LFA-1, Mac-1, and p150,95, respectively. LFA-1 is the receptor for three members of the Ig supergene family of proteins, ICAM-1 (CD54), ICAM-2 (CD102), and Mac-1 and p150,95 bind to ICAM-1, fibrinogen, and iC3b. ICAM-3 (CD50). CD18/CD11 heterodimeric molecules are involved with cell/cell and cell/extracellular adhesion in immune and inflammatory responses. This MAb blocks theseMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ITGB2
Antigen name CD18
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 405S
Clone 68-5A5
Reactivity Human
Host Mouse
Gene ID Unigene ID: 375957
Isotype IgG2a kappa
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR