Anti-HSPG2 Rat Monoclonal Antibody (CF647) [clone: A7L6]

Supplier: Biotium

BNC470315-100 BNC470315-500
75981-042EA 325.12 CAD
75981-042 75981-210
Anti-HSPG2 Rat Monoclonal Antibody (CF647) [clone: A7L6]
Antibodies
This MAb specifically precipitates heterogeneous material of high MW, identified as perlecan, a major heparan-sulfate proteoglycan (HSPG) within all basement membranes and cell surfaces. It does not cross-react with laminin, fibronectin, or dermatran sulfate proteoglycan. Because of perlecan s strategic location and ability to store and protect growth factors, it has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a co-receptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: HSPG2
Clonality: Monoclonal
Clone: Clone A7L6
Conjugation: CF647
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Cow, Fish, Human, Monkey, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR