Anti-CD36 Mouse Monoclonal Antibody [clone: 1E8.]

Supplier: Biotium

BNUB0314-100 BNUB0314-500
75894-980EA 380.65 CAD
75894-980 75894-982
Anti-CD36 Mouse Monoclonal Antibody [clone: 1E8.]
Antibodies
This antibody recognizes a protein of 80 kDa-90 kDa, identified as CD36. It is expressed on platelets, monocytes and macrophages, microvascular endothelial cells, erythrocyte precursors, mammary epithelial cells, and some macrophage derived dendritic cells. CD36 acts as a receptor for thrombospondin (TSP), collagen types I, IV and V, P. falciparum malaria-infected erythrocytes, and sickle erythrocytes. It also functions as a scavenger receptor, mediating macrophage uptake of oxidized low-density lipoprotein (LDL) and recognition of apoptotic polymorphonuclear leukocytes (PMN). CD36 plays aMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD36
Antigen name CD36
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 1E8.
Reactivity Human
Host Mouse
Gene ID Unigene ID: 120949
Isotype IgG1 kappa
ImmunoFluorescence Yes
Concentration 0.2 mg/mL
Storage buffer PBS/0.05% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR