Anti-ITGAL Mouse Monoclonal Antibody (CF770) [clone: DF1524]

Supplier: Biotium

BNC700312-100 BNC700312-500
75886-714EA 325.12 CAD
75886-714 75886-716
Anti-ITGAL Mouse Monoclonal Antibody (CF770) [clone: DF1524]
Antibodies
This antibody recognizes a protein of 180 kDa, identified as CD11a (Leucocyte Workshop IV; Code 1524). CD11a complex with the 2 subunit of the integrin family, CD18, to form the cell surface heterodimer, LFA-1 or CD11a /C18 (aLbL). LFA-1 is expressed on all leukocytes including lymphocytes, monocytes, and granulocytes. It is involved in leukocyte adhesion to its ligands including intercellular adhesion molecule-1 (ICAM-1 or CD54), ICAM-2 (CD102), ICAM-3 (CD50) and Telencephalin (TLN) and play a role in most immune/inflammatory responses. This MAb potently blocks LFA-1 dependent homotypic cell aggregation.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®770 is a near-infrared fluorescent dye (Ex/Em 770/797 nm) with exceptional brightness for in vivo imaging or near-IR western blotting.

Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: Clone DF1524
Conjugation: CF770
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR