Anti-CD48 Mouse Monoclonal Antibody (Biotin) [clone: 156-4H9]

Supplier: Biotium

BNCB0307-100 BNCB0307-500
75911-832EA 325.12 CAD
75911-832 75911-834
Anti-CD48 Mouse Monoclonal Antibody (Biotin) [clone: 156-4H9]
Antibodies
This antibody reacts with human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of marker for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: Clone 156-4H9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR