Anti-IL2RA Mouse Monoclonal Antibody (CF568) [clone: 143-13]

Supplier: Biotium

BNC680297-500 BNC680297-100
75890-968EA 900.17 CAD
75890-968 75891-174
Anti-IL2RA Mouse Monoclonal Antibody (CF568) [clone: 143-13]
Antibodies
This antibody recognizes a protein of 55 kDa, identified as CD25 (Workshop IV; Code A27). CD25 is expressed on activated T- and B-cells and activated monocytes/macrophages. With respect to lymphomas, CD25 is present on malignant cells of Hodgkins disease, HTLV-1 adult T-cell leukemia, cutaneous T-cell lymphoma, and hair cell leukemia. Increased levels of soluble CD25 are observed in the leukemias/lymphomas and inflammatory/ autoimmune diseases. CD25 molecule alone appears to function as a low affinity receptor and associates with CD122 (IL-2R beta chain, p75) and CD132 (common gamma chain) to form the high affinity IL-2 receptor complex. CD25 antibodies detect three epitope regions, A, B and C. This MAb This antibody recognizes the epitope B, which is located at residue 3-104 of CD25 and can effectively block IL-2 binding to CD25.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: Clone 143-13
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR