Anti-NKX2-2 Mouse Monoclonal Antibody [clone: NX2/294]

Supplier: Biotium

BNUB0294-100 BNUB0294-500
75968-586EA 380.65 CAD
75968-586 75968-588
Anti-NKX2-2 Mouse Monoclonal Antibody [clone: NX2/294]
Antibodies
Expression of NKX2.2 has been found in neuroendocrine tumors of the gut, making it a potential marker for the study of gastrointestinal neuroendocrine tumors. More recently, NKX2.2 protein was identified as a target of EWS-FLI-1, the fusion protein specific to Ewing sarcoma, and was shown to be differentially upregulated in Ewing sarcoma on the basis of array-based gene expression analysis. It acts as a valuable marker for Ewing sarcoma, with a sensitivity of 93% and a specificity of 89%, and aids in the differential diagnosis of small round cell tumors.

Type: Primary
Antigen: NKX2-2
Clonality: Monoclonal
Clone: Clone NX2/294
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR