Anti-FUT4 Mouse Monoclonal Antibody [clone: Leu-M1]

Supplier: Biotium

BNUB0277-100 BNUB0277-500
75888-136EA 380.65 CAD
75888-136 75888-138
Anti-FUT4 Mouse Monoclonal Antibody [clone: Leu-M1]
Antibodies
CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkins disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: Clone Leu-M1
Conjugation: Unconjugated
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR