Anti-FN1 Mouse Monoclonal Antibody (CF488A) [clone: HFN7.1]

Supplier: Biotium

BNC880270-100 BNC880270-500
75926-856EA 325.12 CAD
75926-856 75926-858
Anti-FN1 Mouse Monoclonal Antibody (CF488A) [clone: HFN7.1]
Antibodies
Fibronectin is a dimeric glycoprotein of 440 kDa, which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing. This MAb is directed against the peptide core and reacts with both the plasma and cellular forms of fibronectin. It blocks the fibronectin-medicated cell attachment not by disrupting the collagen-fibronectin interaction, but by interfering with the attachment of fibronectin to its receptor on the cell surface.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a green fluorescent dye (Ex/Em 490/515 nm) with excellent brightness and photostability. The dye is minimally charged for less non-specific binding. CF®488A also is compatible with super-resolution imaging by TIRF.

Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: Clone HFN7.1
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR