Anti-SHBG Mouse Monoclonal Antibody [clone: SHBG/245]

Supplier: Biotium

BNUB0245-100 BNUB0245-500
75974-098EA 380.65 CAD
75974-098 75974-100
Anti-SHBG Mouse Monoclonal Antibody [clone: SHBG/245]
Antibodies
This antibody recognizes a protein of 45 kDa, identified as SHBG. It functions as an androgen transport protein, but may also be involved in receptor-mediated processes. Each dimer binds one molecule of steroid. It is specific for 5-alpha-dihydrotestosterone, testosterone, and 17-beta-estradiol. SHBG regulates the plasma metabolic clearance rate of steroid hormones by controlling their plasma concentration. In testis, it is synthesized by the Sertoli cells, secreted into the lumen of the seminiferous tubule and transported to the epididymis.

Type: Primary
Antigen: SHBG
Clonality: Monoclonal
Clone: Clone SHBG/245
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR