Anti-SHBG Mouse Monoclonal Antibody (CF405S) [clone: SHBG/245]

Supplier: Biotium

BNC040245-100 BNC040245-500
75974-116EA 325.12 CAD
75974-116 75974-118
Anti-SHBG Mouse Monoclonal Antibody (CF405S) [clone: SHBG/245]
Antibodies
This antibody recognizes a protein of 45 kDa, identified as SHBG. It functions as an androgen transport protein, but may also be involved in receptor-mediated processes. Each dimer binds one molecule of steroid. It is specific for 5-alpha-dihydrotestosterone, testosterone, and 17-beta-estradiol. SHBG regulates the plasma metabolic clearance rate of steroid hormones by controlling their plasma concentration. In testis, it is synthesized by the Sertoli cells, secreted into the lumen of the seminiferous tubule and transported to the epididymis.

CF® dyes are Biotium's next-generation fluorescentMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol SHBG
Antigen name SHBG
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 405S
Clone SHBG/245
Reactivity Human
Host Mouse
Gene ID Unigene ID: 632235
Isotype IgG1
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR