Anti-MFGE8 Mouse Monoclonal Antibody (Biotin) [clone: MFG-06]

Supplier: Biotium

BNCB0227-100 BNCB0227-500
75961-334EA 325.12 CAD
75961-334 75961-336
Anti-MFGE8 Mouse Monoclonal Antibody (Biotin) [clone: MFG-06]
Antibodies
This antibody recognizes a protein of 40-45 kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.

Type: Primary
Antigen: MFGE8
Clonality: Monoclonal
Clone: Clone MFG-06
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR