Anti-KRT17 Mouse Monoclonal Antibody [clone: E3]
Supplier: Biotium
BNUB0158-100 BNUB0158-500
75916-314EA 380.65 CAD
75916-314 75916-316
Anti-KRT17 Mouse Monoclonal Antibody [clone: E3]
Antibodies
Cytokeratin 17 (CK17) is normally expressed in the basal cells of complex epithelia but not in stratified or simple epithelia. Antibody to CK17 is an excellent tool to distinguish myoepithelial cells from luminal epithelium of various glands such as mammary, sweat and salivary. CK17 is expressed in epithelial cells of various origins, such as bronchial epithelial cells and skin appendages. It may be considered as an epithelial stem cell marker because CK17 Ab marks basal cell differentiation. CK17 is expressed in SCLC much higher than in LADC. Eighty-five percent of the triple negativeMore Product Information