Anti-S100A1 Mouse Monoclonal Antibody (CF568) [clone: 4C4.9]

Supplier: Biotium

BNC680143-100 BNC680143-500
75973-688EA 325.12 CAD
75973-688 75973-690
Anti-S100A1 Mouse Monoclonal Antibody (CF568) [clone: 4C4.9]
Antibodies
S100 belongs to the family of calcium binding proteins. S100A and S100B proteins are two members of the S100 family. S100A is composed of an alpha and a beta chain whereas S100B is composed of two beta chains. This antibody is specific against an epitope located on the beta-chain (i.e. in S-100A and S-100B) but not on the alpha-chain of S-100 (i.e. in S-100A and S100A0). This antibody can be used to localize S-100A and S-100B in various tissue sections. S-100 protein has been found in normal melanocytes, Langerhans cells, histiocytes, chondrocytes, lipocytes, skeletal and cardiac muscle,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol S100A1
Environmentally Preferable
Antigen name S100
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 568
Clone 4C4.9
Reactivity Human,Bovine,Rat,Mouse
Host Mouse
Gene ID Unigene ID: 515715
Isotype IgG2a kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR