Anti-Hepatocyte Specific Antigen Mouse Monoclonal Antibody (Biotin) [clone: HSA133]

Supplier: Biotium

BNCB0133-100 BNCB0133-500
75951-172EA 325.12 CAD
75951-172 75951-174
Anti-Hepatocyte Specific Antigen Mouse Monoclonal Antibody (Biotin) [clone: HSA133]
Antibodies
MAb HSA133 stains human liver canaliculi and a subset of hepatocellular carcinomas. In frozen sections, it stains liver canaliculi strongly and may be used as a marker of this hepatic substructure. Cell preparations of hepatocellular carcinoma biopsies and cell lines are found to bind this MAb on the cell surface. HSA133 strongly stains liver canaliculi and hepatic carcinoma cells using frozen sections or paraformaldehyde fixed cell preparations.

Type: Primary
Antigen: Hepatocyte Specific Antigen
Clonality: Monoclonal
Clone: Clone HSA133
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR