Anti-Lung Specific Antigen Mouse Monoclonal Antibody [clone: LSA120]

Supplier: Biotium

BNUB0120-100 BNUB0120-500
75957-138EA 380.65 CAD
75957-138 75957-140
Anti-Lung Specific Antigen Mouse Monoclonal Antibody [clone: LSA120]
Antibodies
LSA120 MAb is directed against a lung cell antigen. It shows a well-defined reactivity with cells of the lung lineage at all stages of maturation. Non-lung lineages are negative by immuno-histology. Although the exact identity of its antigen has yet to be determined, it has been shown to be distinct from MOC-52 and UM-A9 proteins. This MAb is useful in the identification of normal and neoplastic human lung cells.

Type: Primary
Antigen: Lung Specific Antigen
Clonality: Monoclonal
Clone: Clone LSA120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen name Lung Specific Antigen
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone LSA120
Reactivity Human
Host Mouse
Isotype IgG1 kappa
ImmunoFluorescence Yes
Concentration 0.2 mg/mL
Storage buffer PBS/0.05% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR