Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF543) [clone: 235-1]

Supplier: Biotium

BNC430099-100 BNC430099-500
75951-856EA 325.12 CAD
75951-856 75951-858
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF543) [clone: 235-1]
Antibodies
This MAb is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. MAb 235-1 recognizes an antigen associated with the nuclei in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in subcellular fractions. It produces a speckled pattern in normal and malignant cells and may be used to stain the nuclei of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®543 is an orange fluorescent dye (Ex/Em 541/560 nm) designed for the 543 nm laser line. It yields the brightest conjugates among spectrally similar dyes.

Type: Primary
Antigen: Human Nuclear Antigen
Clonality: Monoclonal
Clone: Clone 235-1
Conjugation: CF543
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Non-human primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR