Anti-Nuclear Membrane Mouse Monoclonal Antibody (Biotin) [clone: NM97]

Supplier: Biotium

BNCB0097-100 BNCB0097-500
75968-676EA 325.12 CAD
75968-676 75968-678
Anti-Nuclear Membrane Mouse Monoclonal Antibody (Biotin) [clone: NM97]
Antibodies
This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the nuclear membrane expressed in human cells. It can be used to stain the nuclear membrane in cell or tissue preparations and can be used as a marker of the nuclear membrane in subcellular fractions. It produces a ring pattern around the nucleus of cells of normal and malignant cells and may be used to stain the nuclear membrane of cells in fixed or frozen tissue sections.

Type: Primary
Antigen: Nuclear Membrane
Clonality: Monoclonal
Clone: Clone NM97
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR