Anti-CR1 Mouse Monoclonal Antibody (CF647) [clone: E11]

Supplier: Biotium

BNC470040-100 BNC470040-500
75894-766EA 325.12 CAD
75894-766 75894-768
Anti-CR1 Mouse Monoclonal Antibody (CF647) [clone: E11]
Antibodies
This antibody recognizes a protein of 210-220 kDa, which is identified as the complement receptor 1 (CR1)/CD35. This MAb is specific for a site in CR1 that is distal from the C3b-binding site, so that it is unable to block CR1 activity. This MAb is highly specific to CR1 and shows no cross-reaction with CR2. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules. The Knops blood group system is a system of antigens located on this protein. Follicular dendriticMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CR1
Environmentally Preferable
Antigen name CD35 / CR1
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 647
Clone E11
Reactivity Baboon,Cynomolgus monkey,Rhesus monkey,Human
Host Mouse
Gene ID Unigene ID: 334019
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR