Anti-IGKC Mouse Monoclonal Antibody (CF568) [clone: L1C1]

Supplier: Biotium

BNC680018-100 BNC680018-500
75955-620EA 325.12 CAD
75955-620 75955-622
Anti-IGKC Mouse Monoclonal Antibody (CF568) [clone: L1C1]
Antibodies
This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of kappa to lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the kappa light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. Demonstration of clonality in lymphoidMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IGKC
Environmentally Preferable
Antigen name Kappa Light Chain
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 568
Clone L1C1
Reactivity Human
Host Mouse
Gene ID Unigene ID: 449609
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR