Anti-TACSTD1 Mouse Monoclonal Antibody (CF594) [clone: VU-1D9]

Supplier: Biotium

BNC940017-100 BNC940017-500
75893-982EA 325.12 CAD
75893-982 75893-984
Anti-TACSTD1 Mouse Monoclonal Antibody (CF594) [clone: VU-1D9]
Antibodies
This antibody reacts with the first EGF repeat in the extracellular domain of Ep-CAM. It is a 40-43 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas with the exception of adult squamous epithelium, hepatocytes and gastric epithelial cells. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: TACSTD1
Clonality: Monoclonal
Clone: Clone VU-1D9
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR