Anti-MLANA Mouse Monoclonal Antibody (CF594) [clone: M2-7C10]

Supplier: Biotium

BNC940009-100 BNC940009-500
75960-384EA 325.12 CAD
75960-384 75960-386
Anti-MLANA Mouse Monoclonal Antibody (CF594) [clone: M2-7C10]
Antibodies
This antibody recognizes a protein doublet of 20-22 kDa, identified as MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum. This MAb labels melanomas and other tumors showing melanocytic differentiation. It is also a useful positive-marker for angiomyolipomas. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: Clone M2-7C10
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Horse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR