Lab Scoops
Supplier: TWD Scientific
ASPS-03 GSS-03-25
76197-156CS 1035.28 CAD
76197-156 76197-158
Lab Scoops
Scoops
Disposable Lab Scoops allow for easy and rapid transfer of milligram quantities. Scoops are available in either anti-static polypropylene or stainless steel construction.
  • Cost effective
  • Reduces time and labor of repetitive weighing
  • Easy dispense packages
These cost-effective scoops are great for rapid transfer of milligram quanities.