Culture Tubes
Supplier: TWD Scientific
DPTT16X150 DPTT18X150 DPTT20X150 DPTT25X150
76197-160CS 295.89 CAD
76197-160 76197-162 76197-164 76197-166
Culture Tubes
Tubes Culture Tubes
Low-cost polypropylene tubes are available in the larger sizes and capacities required for detailed analysis. A lip at the top reinforces the rim and facilitates handling.
  • Disposable polypropylene test tube
  • Extra-long, tall format
  • Lip at the top reinforces the rim and facilitates handling
  • Autoclavable
Multiple sizes available to meet all your needs.