GENESYS™ Visible and UV-Visible Spectrophotometers, Thermo Scientific
Supplier: Thermo Scientific
840-297000 840-298000 840-308000 840-300000 840-309000 840-301000
76176-738EA 8975.6 CAD
76176-738 76176-740 76176-742 76176-744 76176-746 76176-748
GENESYS™ Visible and UV-Visible Spectrophotometers, Thermo Scientific
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Built on the foundation of 60 years of spectroscopy experience, the next generation Thermo Scientific GENESYS family of Vis/UV-Vis spectrophotometers provides exceptional performance with a contemporary design.
  • Start taking measurements immediately with the simple-to-use, high-resolution, 7-inch color-display touchscreen start menu
  • Exchange samples with ease with large, accessible sample compartment
  • Save and print your data fast any way you want it with multiple data-export options
  • GENESYS 140, 150, 180 and BioMate 160 are Wi-Fi ready with optional USB
More Product Information