Bovine Ribonuclease A (from Pancreas)
Supplier: TEKNOVA CA
R3600 R3601
CA100216-926EA 971.53 CAD
CA100216-926 CA100218-566 CA100219-386
Bovine Ribonuclease A (from Pancreas)
Enzymes
Ribonuclease A, from bovine pancreatic is suitable for molecular biology, biochemical, and cell culture applications.
  • Purity >99.0%
  • Salt fractionated and chromatograpically purified
  • Protease- and salt- free