Trypticase Soy Agar Plates, Teknova
Supplier: Teknova
T1144 T1144
CA100218-988PK 841.67 CAD
CA100218-988 100218-988
Trypticase Soy Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
Contains 1.5% tryptone, 0.5% soytone, 0.5% NaCl, and 1.5% agar.
  • 20 plates
  • Sterile
Ambient shipping conditions. Store at room temperature.