Funnels, Separatory, Squibb, PTFE Stopcock, PE Stopper, Chemglass
Supplier: Chemglass
CG-1747-01 CG-1747-02 CG-1747-03 CG-1747-05 CG-1747-06 CG-1747-07 CG-1747-08 CG-1747-09 CG-1747-10 CG-1747-04
89028-692EA 284.48 CAD
89028-692 89028-694 89028-696 89028-700 89028-702 89028-704 89028-706 89028-708 89028-710 89028-698
Funnels, Separatory, Squibb, PTFE Stopcock, PE Stopper, Chemglass
Funnels Separating Funnels
Squibb-style separatory funnel with a PTFE stopcock.
Delivery information: Supplied completely with a flask length PE stopper.