Macro Cuvettes, UV Quartz, Fireflysci
Supplier: FIREFLYSCI, INC.
1UV1 1UV2 1UV5 1UV10 1UV20 1UV30 1UV40 1UV50 1UV100
76015-100EA 443.96 CAD
76015-100 76015-102 76015-104 76015-106 76015-108 76015-110 76015-112 76015-114 76015-116
Macro Cuvettes, UV Quartz, Fireflysci
Cuvettes Spectrophotometer Cuvettes
Standard spectrophotometer cuvette for UV/VIS spectroscopy.
  • Gives highly accurate results, only need to test sample once
  • Comes in UV Quartz materials
  • Excellent transmission values
  • Fire-fused to ensure that cuvettes are robust and durable
  • Two windows polished