Big Blue, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
200221 200220
CA95040-874EA 5712.62 CAD
CA95040-874 CA99900-318 CA99900-322
Big Blue, Agilent Technologies
Assays Cellular Assays
These are used in all Big Blue rodents and cell lines.
Transpack packaging extract is optimized for use with Big Blue Rodents and Cell lines as well as the lambda CII select Assay.
The Big Blue transgenic rodent mutagenesis assay is an in vivo system for assessing the genetic toxicity of test compounds.
The Lambda Select-cII mutation detection system for Big Blue rodents is a positive mutant-screening method for mutagenesis studies using Big Blue transgenic rodents.
Big Blue paque color controls with four separate phage suspensions of color mutants mixed with non-mutant phage.