Koser Citrate Medium, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M069-500G M069-2.5KG M069-5KG
CA61000-002EA 1005.83 CAD
CA61000-002 CA89222-400 CA89222-402
Koser Citrate Medium, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Koser citrate medium is used to differentiate Escherichia coli from Enterobacter aerogenes on the basis of citrate utilization.
  • White to cream colored
  • homogeneous, free flowing powder
  • Reaction of 0.57 w/v aqueous solution at 25 °C
Caution: Store below 30 °C in tightly closed container and prepared medium at 2 to 8 °C.