Standard For Osmometer, Advanced Instruments
Supplier: ADVANCED INSTRUMENTS INC CA
3MA020
3MA040
3MA100
CA89369-182PK
388.95
CAD
CA89369-182
CA89369-184
CA89369-186
Standard For Osmometer, Advanced Instruments
Standards
Osmolality Standards
Calibration standard is used to check the perforance of osmometer and test methods.