HiVeg™ Tryptone Glucose Extract Agar, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV014-500G
MV014-2.5KG
MV014-5KG
CA61000-246EA
1238.75
CAD
CA61000-246
CA89227-196
CA89227-198
HiVeg™ Tryptone Glucose Extract Agar, HiMedia Laboratories
Media for Microbiology
Dehydrated Media for Microbiology
Used for the enumeration of bacteria in water, air, milk and dairy products.
- Light yellow colored
- Homogeneous, free flowing powder
- Reaction of 2.4% w/v aqueous solution at 25 °C
Caution: Store below 30 °C and the prepared medium at 2 to 8 °C.