Veg 8 Juice Broth, Himedia®
Supplier: HIMEDIA LABORATORIES PVT. CA
M895-500G
M895-2.5KG
M895-5KG
CA61000-208EA
1803
CAD
CA61000-208
CA89226-714
CA89226-716
Veg 8 Juice Broth, Himedia®
Media for Microbiology
Dehydrated Media for Microbiology
Veg 8 juice broth is used for cultivation of fungi-yeasts and moulds.
- Off-white to light yellow colored
- Homogeneous, free flowing powder
- Reaction of 2.43% w/v aqueous solution at 25 °C
Caution: Store dehydrated and the prepared medium at 2 to 8 °C in tightly closed container.