Sporulation Broth, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M1018-500G M1018-2.5KG M1018-5KG
CA95020-298EA 1351.9 CAD
CA95020-298 CA89222-608 CA89222-610
Sporulation Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Sporulation broth is used for promoting sporulation in Bacillus subtilis.
  • Cream to light yellow colored
  • Homogeneous, free flowing powder
  • Reaction of 1.58% w/v aqueous solution at 25 °C
Caution: Store below 30 °C in tightly closed container and the prepared medium at 2 to 8 °C.